CHO cell culture platform liquid media, HyClone™ PF-CHO LS and PF-CHO MPS
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH30333.01 SH30333.02 SH30359.02 SH30334.01 SH30335.01
HYCLSH30333.01EA 258 EUR InStock
HYCLSH30333.01 HYCLSH30333.02 HYCLSH30359.02 SH30334.01 SH30335.01
CHO cell culture platform liquid media, HyClone™ PF-CHO LS and PF-CHO MPS
Zellkulturmedien
HyClone™ PF-CHO LS and PF-CHO MPS media are designed to support the DHFR selection/amplification system. They have been successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks, and bioreactors. PF-CHO MPS media is the powder version of PF-CHO LS media.
  • Designed to support the DHFR selection/amplification system
  • Successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks and bioreactors
  • Enable growth of many CHO cells with minimal adaptation
  • PF-CHO LS liquid medium contains 4 mM L-glutamine and 0,1% poloxamer 188
  • PF-CHO MPS powder medium does not contain L-glutamine or poloxamer 188
HyClone™ PF-CHO LS and PF-CHO MPS media are ideally suited for the cost-effective manufacture of recombinant proteins for academic and industrial applications, genomics and proteomics, in vitro diagnostics, drug target screening and validation, and manufacturing of pre-clinical lots.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..