Screw vials, Thermo Scientific™ Chromacol™
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.1-STVGN
HYPE1.1-STVGNEA 553 EUR InStock
HYPE1.1-STVGN
Screw vials, Thermo Scientific™ Chromacol™
Vials
Thermo Scientific™ Chromacol™ sample storage screw thread vials are manufactured from superior quality 33 expansion borosilicate glass, clear (Type 1, Class A).
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge