Solid phase extraction cartridges, HyperSep™ SCX
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60108-520 60108-433 60108-423 60108-716 60108-717 60108-718 60108-857 60108-420 60108-421 60108-422
HYPE60108-520EA 284 EUR InStock
HYPE60108-520 HYPE60108-433 HYPE60108-423 HYPE60108-716 HYPE60108-717 HYPE60108-718 HYPE60108-857 HYPE60108-420 HYPE60108-421 HYPE60108-422
Solid phase extraction cartridges, HyperSep™ SCX
Chromatographiesäulen
A strong cation exchange sorbent for extraction of charged basic compounds.
  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge