Solid phase extraction cartridges, HyperSep™ Aminopropyl
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60108-519 60108-432 60108-518 60108-364 60108-424 60108-425 60108-738 60108-739 60108-740
HYPE60108-519EA 0 EUR InStock
HYPE60108-519 HYPE60108-432 HYPE60108-518 HYPE60108-364 HYPE60108-424 HYPE60108-425 HYPE60108-738 HYPE60108-739 HYPE60108-740
Solid phase extraction cartridges, HyperSep™ Aminopropyl
Chromatographiesäulen
A polar sorbent for both polar and anion exchange interactions.
  • Weak anion exchanger
  • Enhanced retention of strong anions, such as sulfonic acids
  • Excellent retention of drugs, metabolites and structural isomers
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge