Solid phase extraction cartridges, HyperSep™ Cyano
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
60108-745 60108-746 60108-747 60108-748 60108-750 60108-751 60108-752
HYPE60108-745EA 342 EUR InStock
HYPE60108-745 HYPE60108-746 HYPE60108-747 HYPE60108-748 HYPE60108-750 HYPE60108-751 HYPE60108-752
Solid phase extraction cartridges, HyperSep™ Cyano
Chromatographiesäulen
Solid phase extraction cartridges with medium polarity sorbent, ideal for the isolation of polar compounds from non polar matrices.
  • Less retentive than silica or diol
  • Used for retention of polar compounds from hexane and oils
  • Suited for reversed phase extraction of moderately polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Verpackung: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge