SureSTART™ Septa, 22 mm, Level 2 High-throughput Applications
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6A24ST26 6A20ST20
HYPE6A24ST26EA 86.8 EUR InStock
HYPE6A24ST26 HYPE6A20ST20
SureSTART™ Septa, 22 mm, Level 2 High-throughput Applications
Septen
Level 2 septa are manufactured and tested to ensure low bleeding, robustness, and reproducibility of results for your routine GC-MS and LC-MS analyses.
  • Autoclavable
  • Single use
  • Non sterile
Choose all-purpose silicone/PTFE septa manufactured with a shore value of 50 to reduce autosampler needle issues.
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge