SureSTART™ Specification Certified Vial and Cap Kits, Level 1
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6EKSHV8-2
HYPE6EKSHV8-2EA 59.1 EUR InStock
HYPE6EKSHV8-2
SureSTART™ Specification Certified Vial and Cap Kits, Level 1
Vials
Select Level 1 products for an affordable option for everyday chromatography analysis using core detector technology.
  • For everyday analysis
  • Chromatography approved
  • Not autoclavable
  • No preservative
  • Shell vials
Choose from a variety of kits containing short thread screw vials in clear propylene and polypropylene caps with silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues.
SureSTART Level 1 products are cost optimised, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge