Glass screw top vials, 2 ml, level 3 high performance applications, SureSTART™
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6PSV9-1P 6PSV9-1PF 6PSV9-1PSS 6PSV9-1PSSF 6PSV9-1PSST 6PSV9-1PT 6PSV9-2PSS 6PSV9-2PSSF 6PSV9-2PSST 6PSV9-S1P
HYPE6PSV9-1PEA 19.8 EUR InStock
HYPE6PSV9-1P HYPE6PSV9-1PF 548-2773 HYPE6PSV9-1PSSF HYPE6PSV9-1PSST HYPE6PSV9-1PT HYPE6PSV9-2PSS HYPE6PSV9-2PSSF HYPE6PSV9-2PSST HYPE6PSV9-S1P
Glass screw top vials, 2 ml, level 3 high performance applications, SureSTART™
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml GOLD-grade clear glass screw and crimp top vials, performance level 3, when analysing analytes such as polar compounds that strongly adsorb on the glass surface or are sensitive to sample pH changes.
Level 3 vials are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. This offering includes Thermo Scientific™ SureStop™ vials, which, when used with 9 mm AVCS™ screw caps, ensure an optimal seal every time. The SureStop vial-AVCS cap system is specially designed to prevent screw thread vial failures such as cap tilt, cap deformities, dislodged septa, or septum push through, thus preventing injection failures and sample evaporation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge