PTFE/silicone discs, Tuf-Bond™
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TS-12712 TS-12708 TS-12713 TS-12716 TS-12718 TS-12720 TS-12722
HYPETS-12712EA 117 EUR InStock
HYPETS-12712 HYPETS-12708 HYPETS-12713 HYPETS-12716 HYPETS-12718 HYPETS-12720 HYPETS-12722
PTFE/silicone discs, Tuf-Bond™
Septen
These discs ensure air-tight, inert seals. They are specifically designed to combine the resealability of silicone with the inertness of PTFE.
  • Easily penetrated with standard syringe and CC needles
  • PTFE layer is 0,25 mm thick to ensure protection
These discs reseal instantly puncture after puncture, and the PTFE coating is structurally bonded so that no cement is leached or baked out of your sample after needle penetration.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge