Small reaction vials, Reacti-Vial™
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TS-13225 TS-13100 TS-13220 TS-13221 TS-13097 TS-13223 TS-13222 TS-13099
HYPETS-13225EA 441 EUR InStock
HYPETS-13225 HYPETS-13100 HYPETS-13220 HYPETS-13221 HYPETS-13097 HYPETS-13223 HYPETS-13222 HYPETS-13099
Small reaction vials, Reacti-Vial™
Vials
These small reaction vials have an internal cone designed to make small-sample collection and handling easy and convenient. The cone feature is particularly useful for removing small quantities of sample with a syringe.
  • Extra thick glass wall magnifies the sample ideal for observing chemical reactions
  • Amber vials are light-sensitive to well protect the compounds
These vials are ideal for residue isolation, derivative preparation, maximum sample retrieval, moisture protection, sample storage, purification, precipitations, centrifugations and solvent separations.
Lieferumfang: Vials are supplied complete with open-top screw caps and teflon/rubber laminated discs.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge