Immersion oil viscosity 150 cSt (lit.), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
I0765-100ML I0765-500ML
I0765-100MLEA 69.5 EUR InStock
I0765-100ML I0765-500ML
Immersion oil viscosity 150 cSt (lit.), Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
Immersion Oil has a defined refractive index of about 1,516. It is important that the refractive index (nD) is about 1,5, similar to that of glass. This enables a homogeneous oil immersion to be achieved.
Immersion media are used in conjunction with immersion objectives and are located between the surface of the specimen and the lens of the microscope. Immersion oils practically eliminate light beam deflection so that the effectiveness of the lens is considerably increased.
Immersion oil has a larger impact on 3D resolution. Specifically, matching the immersion oil refractive index to glass, reliably enhances the ability to visualise dendritic spines.
Application:
In epifluorescent microscopy and cell identification.
In the assembly and filling of the microinjecting system.
In microscopy to reliably visualise dendritic spines.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge