Immersion oil viscosity 1,250 cSt (lit.), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
I0890-100ML I0890-500ML
I0890-100MLEA 64.5 EUR InStock
I0890-100ML I0890-500ML
Immersion oil viscosity 1,250 cSt (lit.), Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
Immersion oil has been used in epifluorescent microscopy and cell identification.
  • Suitable for Hematology
  • Suitable for Histology
Immersion oil has a larger impact on 3D resolution. Specifically, matching the immersion oil refractive index to glass, reliably enhances the ability to visualize dendritic spines.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge