Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid kolorimetrische Tests, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

I8377-1G I8377-250MG I8377-25G I8377-5G
I8377-1GEA 273 EUR InStock
I8377-1G I8377-250MG I8377-25G I8377-5G
2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid kolorimetrische Tests, Sigma-Aldrich®
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Iodonitrotetrazolium chloride has been used for the staining of cells. It has been used as a coupling agent in the enzymatic assay of D-arabinitol.

  • Electron acceptor for the colorimetric assay of various dehydrogenases


Formel: C₁₉H₁₃ClIN₅O₂
Melting Pt: 227…229 °C
Storage Temperature: Kühlschrank
MDL Number: MFCD00011961
CAS-Nummer: 146-68-9
EINECS: 205-676-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01c93a5a9d0459e175eafa9cc56edc91a93541eb&vid=01c93a5a9d0459e175eafa9cc56edc91a93541eb&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Light Yellow to Yellow
Appearance (Form) Powder
Solubility (Colour) Faint Green to Light Green and Faint Green-Yellow to Green-Yellow and Light Yellow to Yellow
Solubility (Turbidity)
4 mg/ml, H₂O
Clear
Solvent Content (by GC) ≤4 %
Purity (TLC) ≥98 %
Recommended Retest Period 5 years

 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge