Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Asparagin, wasserfrei ≥99%, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0210079425 0210079480 0210079490 0210079491
ICNA0210079425EA 26.7 EUR InStock
ICNA0210079425 ICNA0210079480 ICNA0210079490 ICNA0210079491
L(+)-Asparagin, wasserfrei ≥99%, weißes Pulver
L(+)-Asparagin
L-Asparagine is used in cell culture media and is a component of MEM non-essential amino acids solution. L-Asparagine has been shown to enhance ornithine decarboxylase activity in cultured human colon adenocarcinoma Caco-2 cells and in cultured IEC-6 intestinal epithelial cells. Spore germination in Bacillus subtilis has been increased in the presence of L-asparagine.

Soluble in aqueous solution (1% - clear, colorless solution).

Achtung! Bitte beachten: Research Use Only (RUO).


Formel: H₂NCOCH₂CH(NH₂)CO₂H
Storage Temperature: Raumtemperatur
MDL Number: MFCD00064401
CAS-Nummer: 70-47-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Loss on Drying ≤1,0%
Optical Rotation (c=10, 1N HCl) +25° to +36°

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge