Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

5-Phospho-D-ribose 1-diphosphate pentasodium salt, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0210098180 0210098110 0210098125
ICNA0210098180EA 1472 EUR InStock
ICNA0210098180 ICNA0210098110 ICNA0210098125
5-Phospho-D-ribose 1-diphosphate pentasodium salt, weißes Pulver
5-Phospho-D-ribose 1-diphosphate pentasodium salt
5-Phosphorylribose-1-pyrophosphate Sodium Salt Hydrate has been used in a study to assess the relationship between the muscarinic receptor cationic current and internal calcium in guinea-pig jejunal smooth muscle cells. It has also been used in a study to investigate inhibitors of the bacterial cell wall biosynthesis of the enzyme Mur D.

Used in a study to assess the relationship between the muscarinic receptor cationic current and internal calcium in guinea-pig jejunal smooth muscle cells.

Soluble in water (50 mg/ml - clear to slightly hazy yellowish solution).


Formel: C₅H₈Na₅O₁₄P₃
MW: 499.98 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00084655
CAS-Nummer: 108321-05-7
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Water Content Report result
Sodium Content Report result
Expiration Date Month/Day/Year

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge