Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

D(+)-Cellobiose ≥99%, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0210129880 0210129890 0210129805 0210129810 0210129825
ICNA0210129880EA 465 EUR InStock
ICNA0210129880 ICNA0210129890 ICNA0210129805 ICNA0210129810 ICNA0210129825
D(+)-Cellobiose ≥99%, weißes Pulver
D(+)-Cellobiose
Cellobiose is a disaccharide composed of two glucose molecules. These are β-1,4-glycosidically linked. It is reducing sugar, which is readily soluble in polar solvents such as water and ethanol. D-Cellobiose is a substrate of β-glucosidase.

  • Refractive Index: 1.65 (Predicted)
  • pKa: 12.39

Cell Culture media supplement. D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

Soluble in water 4% (clear colorless solution); insoluble in alcohol and ether.


Formel: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Siedepunkt: 667.93 °C (760 mmHg) (Lit.)
Melting Pt: 229…239 °C
Dichte: 1.53 g/cm³ (25 °C) (Lit.)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00136034
CAS-Nummer: 528-50-7
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standards
Moisture ≤1.0%
Optical Rotation (c=4, water) +34° ± 3°

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge