Corn Syrup, Liquid, MP Biomedicals
Lieferant: MP Biomedicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0210141301
ICNA0210141301EA 89.8 EUR InStock
ICNA0210141301
Corn Syrup, Liquid, MP Biomedicals
Tierbedarf Tiernahrung
Corn-syrup is a dual acid-enzyme conversion syrup which contains approximately three times as much maltose as regular corn syrups processed to the same degree of conversion using acid only.
  • Conversion: acid-enzyme
  • Maltose Content: ~30%
Corn syrup is used in commercially prepared foods as a thickener, a sweetener and as a humectant - an ingredient that retains moisture and thus maintains a food's freshness.
Achtung! Bitte beachten: Research Use Only (RUO).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge