Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

6-Aminocapronsäure, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0210164591 0210164525 0210164580 0210164590
ICNA0210164591EA 616 EUR InStock
ICNA0210164591 ICNA0210164525 ICNA0210164580 ICNA0210164590
6-Aminocapronsäure, weißes Pulver
6-Aminocapronsäure
ε-Amino-n-caproic acid is a plasmin inhibitor. It inhibits carboxypeptidase B. It promotes rapid dissociation of plasmin by inhibiting the activation of plasminogen. As an inhibitor of plasmin it is has been utilised in the clotting buffer for fibrinogen assays.

EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carbo­xy­peptidase, plasmin, and plasminogen activator. Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

Soluble in water (50 mg/ml); sparingly soluble in alcohol and methanol; practically insoluble in ether; insoluble in ethanol.

Achtung! Bitte beachten: Research Use Only (RUO).


Formel: C₆H₁₃NO₂
MW: 131.17 g/mol
Siedepunkt: 257
Melting Pt: 127
Dichte: 1.48 g/mL (Lit.)
Flash-Pt: 207
Storage Temperature: Raumtemperatur
MDL Number: MFCD00008238
CAS-Nummer: 60-32-2
Merck Index: 13,00431
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
Melting Point 205 - 215 °C

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge