Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

3-Indolessigsäure ~99, weißgraues Puder

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0210203705 0210203725 0210203780
ICNA0210203705EA 23.9 EUR InStock
ICNA0210203705 ICNA0210203725 ICNA0210203780
3-Indolessigsäure ~99, weißgraues Puder
3-Indolessigsäure
Indole-3-acetic acid (IAA) is the most common, naturally-occurring, plant growth regulator - as a principal auxin of higher plants. It is the best known of the auxins, and has been the subject of extensive studies by plant physiologists. Chemically, IAA is a carboxylic acid in which the carboxyl group is attached through a methylene group to the C-3 position of an indole ring.

Indole-3-acetic acid, one of its derivatives bis-2-indolyl disulphide has been used to synthesize tyrosin kinase inhibitors. It can also be used as a starting material to synthesize DBI receptor ligands. Methyl ester of indole-3-acetic acid reacts with dimethylamine and the resulting amide on reduction with Lithium aluminium hydride gives the hallucinogen N,N-Dimethyltryptamine. Used as a plant growth regulator as a principle auxin of higher plants. Can also be used as a starting material to synthesize DBI receptor ligands.

Soluble in methanol, ethanol (5%), Sparingly soluble in water, chloroform. Soluble in acetone, ether and freely soluble in alcohol.


Formel: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00005636
CAS-Nummer: 87-51-4
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge