Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Inosine 5'-monophosphate disodium salt hydrate ≥98%, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0210205005 0210205010 0210205025 0210205080
ICNA0210205005EA 44 EUR InStock
ICNA0210205005 ICNA0210205010 ICNA0210205025 ICNA0210205080
Inosine 5'-monophosphate disodium salt hydrate ≥98%, weißes Pulver
Inosine 5'-monophosphate disodium salt hydrate
IMP a nucleotide produced by the deamination of adenosine monophosphate (AMP); it is the precursor of AMP and GMP in purine biosynthesis and an intermediate in purine salvage and in purine degradation.

Inosine 5′-monophosphate (IMP) is used as a substrate to study the distribution, specificity and kinetics of inosine-5′-monophosphate dehydrogenase (IMPDH).

Inosine monophosphate, or inosinic acid, is the ribonucleotide of hypoxanthine and the first nucleotide formed during purine synthesis.

Soluble in water (5% aq soln).

Achtung! Bitte beachten: Research Use Only (RUO).


Formel: C₁₀H₁₁N₄Na₂O₈P·xH₂O
Storage Temperature: Gefrierschrank
MDL Number: MFCD00150372
CAS-Nummer: 4691-65-0
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
UV/VIS: λ max 248 ± 2 nm
OD250/OD260 1.7 ± 0.2
OD280/OD260 0.3 ± 0.2
Moisture Repoer result
Solubility (5% aq soln) Passes

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge