Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Lecithin aus Eigelb 65 - 75%

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0210214625 0210214680
ICNA0210214625EA 94.8 EUR InStock
ICNA0210214625 ICNA0210214680
Lecithin aus Eigelb 65 - 75%
Lecithin aus Eigelb
Lecithin is the major membrane phospholipid in eukaryotic cells. In addition to being the major structural component of cellular membranes, it serves as a reservoir for several lipid messengers. It is the source of the bioactive lipids lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, plateletactivating factor, and arachidonic acid. Leithin is used for preparation of vesicle, suspensions commonly called liposomes or as monolayers. A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter. Soluble in fat, paraffin, ether, ethanol, chlorinated hydrocarbons. Dispersible in water.


Storage Temperature: Raumtemperatur
MDL Number: MFCD00131449
CAS-Nummer: 93685-90-6
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge