Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Polygalacturonic acid (aus Pektin) ≥86%, Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0210271105 0210271125 0210271180 0210271190
ICNA0210271105EA 64 EUR InStock
ICNA0210271105 ICNA0210271125 ICNA0210271180 ICNA0210271190
Polygalacturonic acid (aus Pektin) ≥86%, Pulver
Polygalacturonic acid
Polygalacturonic acid is a water insoluble, transparent gelatinous acid existing in ripe fruit and some vegetables. It is a product of pectin degradation in plants, is produced via the interaction between pectinase and pectin.

Polygalacturonic acid sequesters metal ions. It also produces gels in the presence of Ca2+ that can be used for drug release or to immobilize cells or enzymes for the production of biomolecules.

In land plants, polygalacturonic acids are major components of cell wall polysaccharides (pectins). Bound polygalacturonase-inhibiting peptides (PGIP) protect pectin from degradation by fungal polygalacturonases.

Soluble in 0.1 N HCI (50 mg/2 ml-clear, colorless solution); insoluble in water and ethanol.


Storage Temperature: Raumtemperatur
MDL Number: MFCD00131972
CAS-Nummer: 25990-10-7
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥86%
FTIR Conforms to Reference
Moisture Content <10%
pH (1% susp.) 2.5 - 3.5
Ash Content ≤2%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge