Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Natriumcholat ≥98,5%, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


02102897.5 0210289705 02102897.1 02102897.3 0210289701
ICNA02102897.5EA 636 EUR InStock
ICNA02102897.5 ICNA0210289705 ICNA02102897.1 ICNA02102897.3 ICNA0210289701
Natriumcholat ≥98,5%, weißes Pulver
Natriumcholat
Cholic acid is a biochemical solubilizing agent (a non-ionic, non-denaturing detergent). Occurs inconjugation with glycine or taurine in bile of most vertebrates.

  • Critical Micelle Concentration (CMC): 14mM

Cholic acid is used in the solubilization of membrane-bound proteins in their native state. It is a component for the preparation of liposomes. It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
Cholate is a bile acid produced in liver during cholesterol breakdown. Cholate downregulates cholesterol-7-α-hydroxylase (rate-limiting step in bile acid synthesis). Cholate, an anionic detergent, is used alone and in combination with urea to extract and reconstitute membrane proteins and protein complexes.

Cholic acid is a biochemical solubilising agent (a non-ionic, non-denaturing detergent). Occurs inconjugation with glycine or taurine in bile of most vertebrates.

Soluble in water (100 mg/ml). Soluble in ethanol (40 mg/ml - clear, colorless solution).


Formel: C₂₄H₃₉NaO₅
MW: 430.56 g/mol
Melting Pt: 198
Storage Temperature: Raumtemperatur
MDL Number: MFCD00150749
CAS-Nummer: 361-09-1
EINECS: 206-643-5
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity (HPLC) ≥98,5%
Loss on Drying ≤5%
pH (2% aq soln) 7,5 - 9,0
Solubility (2% aq soln) Clear and Colorless
Sodium Deoxycholate ≤1,5%
Heavy Metals ≤50 ppm

 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge