Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Tricain-Methansulfonat, Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0210310683 0210310680 0210310605 0210310625
ICNA0210310683EA 919 EUR InStock
ICNA0210310683 ICNA0210310680 ICNA0210310605 ICNA0210310625
Tricain-Methansulfonat, Pulver
Tricain-Methansulfonat
Ethyl-m-Aminobenzoate is a derivative of an isomer of benzocaine.

Ethyl-m-Aminobenzoate is mainly used as an anesthetic in fish, sharks, frogs, toads, salamanders, newts and worms. Concentrations generally used in fish are 25 to 300 mg/L water, with larger fish requiring higher concentrations. Bufo marimus frogs require approximately 1000 mg/L , while Rana pipens, R. temporaria and Xenopus leavis require approximately 300 mg/L for adequate anesthesia.

Ethyl-m-Aminobenzoate is a derivative of an isomer of benzocaine.

Soluble in water (1 g/ml).

Achtung! Bitte beachten: Research Use Only (RUO). Please observe the legal regulations of your country.
Keep away from sunlight, as sunlight will cause degradation.


Formel: C₁₀H₁₅NO₅S
MW: 261.29 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00013176
CAS-Nummer: 886-86-2
EINECS: 212-956-8
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
Melting Point 145 - 150°C
pH (1% aq soln) <7
Loss on Drying ≤5%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge