Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Tributyrin (Glycerintributyrat), Flüssigkeit

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0210311191 0210311180 0210311183 0210311190
ICNA0210311191EA 417 EUR InStock
ICNA0210311191 ICNA0210311180 ICNA0210311183 ICNA0210311190
Tributyrin (Glycerintributyrat), Flüssigkeit
Tributyrin (Glycerintributyrat)
Tributyrin, is a glycerol substituted with three butyrl esters.

Tributyrin induces differentiation of malignant cells followed by arrest of cell growth and apoptosis. It is used as a deliverable source of natural butyrate and is demonstrated to inhibit growth of and induce cell differentiation of Caco-2 cells more potently than natural butyrate.

Tributyrin induces differentiation of malignant cells followed by arrest of cell growth and apoptosis. It is used as a deliverable source of natural butyrate and is demonstrated to inhibit growth of and induce cell differentiation of Caco-2 cells more potently than natural butyrate.

Soluble in alcohol and ether; insoluble in water.


Formel: C₁₅H₂₆O₆
Siedepunkt: 287…288 °C (Lit.)
Melting Pt: −75 °C (Lit.)
Dichte: 1,035 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00009392
CAS-Nummer: 60-01-5
EINECS: 200-451-5
Merck Index: 12,09750
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge