Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Vitamin A acetat, ~500,000 IU/gm, gelbes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0210325710 0210325780
ICNA0210325710EA 30.2 EUR InStock
ICNA0210325710 ICNA0210325780
Vitamin A acetat, ~500,000 IU/gm, gelbes Pulver
alle-trans-Retinylacetat
The product contains vitamin A acetate (approximately 21%) finely dispersed in a starch-coated matrix of gelatin (approximately 60%), sucrose (approximately 14%) and corn starch (approximately 5%). BHT and BHA are added as antioxidants.

  • Concentration: Vitamin A Content: >500,000 IU/g
  • Specific Activity : ~500,000 IU/gm
  • Presentation : Yellow Powder
  • Particle Size : 100% through sieve No. 2; minimum 90% through sieve No. 40; Maximum 15% through sieve No. 100.
  • Loss on Drying : ≤8%

The product contains vitamin A acetate (approximately 21%) finely dispersed in a starch-coated matrix of gelatin (approximately 60%), sucrose (approximately 14%) and corn starch (approximately 5%). BHT and BHA are added as antioxidants.

Vitamin A acetate induces cell differentiation and inhibits cell proliferation.


Formel: C₂₂H₃₂O₂
Melting Pt: 57,..58 °C (Lit.)
Storage Temperature: Kühlschrank
MDL Number: MFCD00019413
CAS-Nummer: 127-47-9
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Loss on Drying <8%
Vitamin A Content >500,000 IU/g
Microbiological Purity Satisfactory

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge