Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin) ≥97%, farbloser Feststoff

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

02152070.5 0215207001 0215207005
ICNA02152070.5EA 51.9 EUR InStock
ICNA02152070.5 ICNA0215207001 ICNA0215207005
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin) ≥97%, farbloser Feststoff
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin)
Spermine is a polyamine involved in cellular metabolism found in all eukaryotic cells. Formed from spermidine through the addition of aminopropyl group to spermidine by spermine synthase. It is found as a polycation at physiological pH.

Mixed NMDA glutamate receptor agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).

Soluble in water (1% w/v), lower alcohol, chloroform; practically insoluble in ether, benzene, petroleum ether.

Achtung! Bitte beachten: Research Use Only (RUO). Desiccate, store under nitrogen.


Formel: C₁₀H₂₆N₄
Siedepunkt: 141…142 °C (0,5 mmHg)
Melting Pt: 28…30 °C (Lit.)
Storage Temperature: Kühlschrank
MDL Number: MFCD00008215
CAS-Nummer: 71-44-3
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥97%
FTIR Conforms to Standard
pH (0.5% aq soln) 11 - 14

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge