Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sudan III, rötlich braunes Pulver C.I. 26100

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0215208725 0215208780
ICNA0215208725EA 107 EUR InStock
ICNA0215208725 ICNA0215208780
Sudan III, rötlich braunes Pulver C.I. 26100
Sudan III
Sudan III is used in Coloring oils, sprit, lacquers etc; also as stain for zoological, pathological and vegetable objects like wax, cutin, resin, contents of lactiferous ducts etc, which are colored red, while cellulose membranes remain uncolored. Approved by FDA for external use only.

Insoluble in water; soluble in chloroform, glacial acetic acid; moderately soluble in alcohol, ether, acetone, fixed and volatile oils, hot glycerol. Clear red solution at 1 mg/ml in toluence.

Achtung! Bitte beachten: Research Use Only (RUO).


Formel: C₂₂H₁₆N₄O
Melting Pt: 195 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003905
CAS-Nummer: 85-86-9
Order Now
Spezifikation Testergebnisse
Identity Test Passes
UV/Vis: λmax (acetone) 502 - 510 nm

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge