Cellulase (from Trichoderma virde), MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0215233705
ICNA0215233705EA 1388 EUR InStock
ICNA0215233705
Cellulase (from Trichoderma virde), MP Biomedicals
Enzyme
Readily soluble in water.
One unit will liberate one µmole of glucose from cellulose in 1 hr at pH 5 and 37 °C. ; CMC Unit Definition: 100 units is sufficient to produce reducing sugar equivalent to 1 mg of glucose per minute at pH 5,0 and 40 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge