Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Hydroxypropyl-β-cyclodextrin, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0215354001 0215354005
ICNA0215354001EA 35.9 EUR InStock
ICNA0215354001 ICNA0215354005
Hydroxypropyl-β-cyclodextrin, weißes Pulver
Hydroxypropyl-β-cyclodextrin
2-Hydroxypropyl-beta-cyclodextrin are non-toxic solubilisers. The solubility of drugs increases linearly with the concentration of 2-Hydroxypropyl-b-cyclodextrin in aqueous buffer. The formation of drug/cyclodextrin complexes is a rapidly reversible reaction and complexes exist both in solution and crystalline states.

Solutions of many such complexes may be lyophilised to produce freely soluble powders which may be compressed into tablets. Bio-effects are only slightly affected by cyclodextrin complexation. Cells in serum supplemented medium can be grown in up to 1 to 2%. In serum-free medium, concentrations of 0,5 to 1% are acceptable. Hydroxypropyl-b-cyclodextrin has been found to be non-toxic in mice and rabbits. 2-hydroxypropyl-g-cyclodextrin with its 8 glucose units, has a slightly larger cavity and can accomodate larger substrates than the beta form. The use of cyclodextrins in receptor binding assays in not recommended.

Soluble in water (50 g/100 ml). Solutions may be obtained by stirring 30 minutes at room temperature. Alternatively, sonication with cooling may be employed. Solutions may be stored for several weeks at room temperature. Good solubility in methanol, ehtanol, pyridine, dimethyl sulphoxide, dimethyl formamide.


Formel: C₅₄H₁₀₂O₃₉
MDL Number: MFCD16621721
CAS-Nummer: 94035-02-6
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge