Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Indocyaningrün ~90%

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0215502025 0215502050 0215502080
ICNA0215502025EA 77.9 EUR InStock
ICNA0215502025 ICNA0215502050 ICNA0215502080
Indocyaningrün ~90%
Indocyanine green
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.

  • Extinction Coefficient (E1%): 10.7-11.6 (393-394 nm)(Lit.)

It is used for determining cardiac output, hepatic function, and liver blood flow, and for ophthalmic angiography. ICG binds tightly to plasma proteins and becomes confined to the vascular system.
Indocyanine Green is a negatively charged polymethine dye that forms noncovalent fluorescent complexes with proteins. This "pseudofluorogenic" property has been utilized for protein determination by capillary electrophoresis with diode laser-induced fluorescence detection. ICG is a fluorescent dye which is used in medicine as an indicator substance (e.g. for photometric hepatic function diagnostics and fluorescence angiography) in cardiac, circulatory, hepatic and ophthalmic conditions. It has been used to resolve several labeled proteins including: human serum albumin, ribonuclease A, transferrin, and cytochrome c.

Sparingly soluble in acetone/glycil ethers and ehtanol; water (10 mg/1mL - transparent solution.)


Formel: C₄₃H₄₇N₂NaO₆S₂
Storage Temperature: Raumtemperatur
MDL Number: MFCD00013078
CAS-Nummer: 3599-32-4
EINECS: 222-751-5
Order Now
Spezifikation Testergebnisse
Identity Test Passes

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge