Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Isoamylalkohol ≥99%, klare, farblose Flüssigkeit

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

0215507880 0215507890 0215507891
ICNA0215507880EA 32.7 EUR InStock
ICNA0215507880 ICNA0215507890 ICNA0215507891
Isoamylalkohol ≥99%, klare, farblose Flüssigkeit
Isoamylalkohol
Isoamy alcohol is routinely used in molecular biology, especially in the purification of DNA. It is widely used in conjunction with phenol and chloroform (25:24:1 phenol : chloroform : isoamyl alcohol) for the removal of proteins from the nucleic acid solutions by extraction.

  • Japan fire code: 4.2
  • One of several isomers of amyl alcohol

Isoamyl alcohol is added to the phenol solution to help inhibit RNase activity and to help prevent the solubilisation in the phenol phase of long RNA molecules with long poly(A) portions. It will also help in reducing the foaming during the extraction process.Isoamyl alcohol is also used in the extraction of ethidium bromide from DNA solutions and in the radiolabeling of RNA transcripts in nuclei that have been isolated from tissue. Isoamy alcohol has various miscellaneous uses which include HPLC analysis of various pharmaceuticals and metabolites, nalysis of oxidised and reduced pyridine nucleotides and adenylates in organic phenol extracts from mitochondria, isolation of mRNA from a thermophilic cyanobacterium, Solvent for fats, resins, alkaloids, Manufacturing isoamyl (amyl) compounds, isovaleric acid, mercury fulminate, pyroxylin, artificial silk, lacquers, smokeless powders, for dehydrating celloidin solutions, for determining fat in milk etc.

Miscible with alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, oils.


Formel: (CH₃)₂CHCH₂CH₂OH
Melting Pt: –179 °F (Lit.)
MDL Number: MFCD00002934
CAS-Nummer: 123-51-3
UN: 1105
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=60e27777862a1441bdec89c868acf27847ec49b8&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge

Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.

Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..