Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

(R)-(+)-Limonen, Flüssigkeit

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0215523480 0215523490
ICNA0215523480EA 47.5 EUR InStock
ICNA0215523480 ICNA0215523490
(R)-(+)-Limonen, Flüssigkeit
(R)-(+)-Limonen
D-Limonene is a chiral intermediate for natural product synthesis. It is used as a renewably based solvent in cleaning products.

D-Limonene is a monoterpene found to prevent mammary cancer by inducing hepatic glutathione-S- transferase and uridine diphosphoglucuronosyl transferase. D-Limonene is an isoprenylation inhibitor of small G-proteins.

Limonene is a cyclie terpene from Chinese medicinal herb essential oils used in the synthesis of carvone. Limonene may be used as a shrinking agent to dissolve polystyrene. Limonene may be used in various insecticidal and insect repellant applications.

Miscible with alcohol, with most fixed oils, mineral oil; very slightly soluble in glycerin; practically insoluble in water and in propyiene glycol.


Formel: C₁₀H₁₆
MW: 136.24 g/mol
Siedepunkt: 175.5
Melting Pt: -74
Dichte: 0.84 g/ml
Flash-Pt: 43
Storage Temperature: Raumtemperatur
MDL Number: MFCD00062991
CAS-Nummer: 5989-27-5
UN: 2052
ADR: 3 III
Merck Index: 13,05515
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge