Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Morin, hellbraunes Pulver C.I. 75660

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0215573305 0215573310
ICNA0215573305EA 55.2 EUR InStock
ICNA0215573305 ICNA0215573310
Morin, hellbraunes Pulver C.I. 75660
Morin
Morin is a flavonoid with anti-oxidant properties.

It has been shown to protect cells against oxygen radical damage. Morin not only scavenges oxyradicals, but also moderately inhibits xanthine oxidase, a free-radical generating enzyme. At concentrations of 75-100 micromolar, morin inhibits oxidation of low density lipoprotein (LDL) by free radicals or Cu2+. Morin has been reported to inhibit rat brain phospha­tidyl­inositol­phos­phate kinase activity in vitro and in vivo.

Soluble in ethanol, methanol (0,5% w/v- clear yellow to brown solution) It is also soluble in water (0,25 mg/ml, 20 °C; 0,94 mg/ml, 100 °C), freely soluble in alcohol, slightly soluble in ether and acetic acid, and soluble in aqueous alkaline solutions.


Formel: C₁₅H₁₀O₇
Melting Pt: 303…304 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00006826
CAS-Nummer: 480-16-0
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge