Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

(-)-Chinin Hydrochlorid Dihydrat, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0215648650 0215648605 0215648625 0215648680
ICNA0215648650EA 161 EUR InStock
ICNA0215648650 ICNA0215648605 ICNA0215648625 ICNA0215648680
(-)-Chinin Hydrochlorid Dihydrat, weißes Pulver
(-)-Chinin Hydrochlorid Dihydrat
Quinine Hydrochloride is an alkaloid originally isolated from the Cinchona family of South American trees. It is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent.

Quinine is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent; also used as a resolving agent. A potassium channel blocker. Also used as a flavoring agent in carbonated beverages, a skeletal muscle relaxant and an antiprotozoal agent for fish. A chiral catalyst used in highly enantioselective [2+2] cycloadditions. A resolving agent for carboxylic acids. Catalyzes the kinetic resolution of furanones.

Potassium channel blocker.

Soluble in water (1 g/16 ml at 25 °C; 1 g/0,5 ml at 100 °C), ethanol (1 g/1 ml), glycerol (~1 g/7 ml), chloroform (~1 g/ml), or ether (1 g/350 ml).


Formel: C₂₀H₂₅ClN₂O₂·2H₂O
MW: 396.91 g/mol
Melting Pt: 115…116 °C (Lit.)
Flash-Pt: 122
Storage Temperature: Raumtemperatur
MDL Number: MFCD00151248
CAS-Nummer: 6119-47-7
EINECS: 205-001-1
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge