Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sudanblau II, dunkel rot/violett fast schwarzes Pulver C.I. 61554

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0215663550 0215663580
ICNA0215663550EA 118 EUR InStock
ICNA0215663550 ICNA0215663580
Sudanblau II, dunkel rot/violett fast schwarzes Pulver C.I. 61554
Sudanblau II
Solvent Blue 35 is a member of the Sudan family of hydrophobic fat-staining dyes. In structural classification it is a diazo dyes.
Solvent Blue 35 is predominantly used for staining triglycerides in animal tissues (frozen sections). With the use of certain solvents, may also be used to stain some protein bound lipids in paraffin sections.

  • Presentation: Dark Red/Purple almost Black Powder
  • Dye content: ∼ 90%
  • Soluble in chloroform(4mg/mL).
  • UV/Visible Absorbance: λmax: 604 nm (2nd), λmax: 652 nm


Formel: C₂₂H₂₆N₂O₂
Melting Pt: 120…122 °C (Lit.)
Dichte: 1,23 g/mL (Lit.)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011714
CAS-Nummer: 17354-14-2
Order Now
Spezifikation Testergebnisse
Identity Test Passes

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge