Sorbitol dehydrogenase, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0215664205
ICNA0215664205EA 613 EUR InStock
ICNA0215664205
Sorbitol dehydrogenase, MP Biomedicals
Enzyme
Sorbitol dehydrogenase has been used in a study to investigate osmotic stress induced oxidative damage as a possible mechanism of cataract formation in diabetes.
Sorbitol dehydrogenase is an oxidoreductase. Sorbitol dehydrogenase is an enzyme in carbohydrate metabolism converting fructose into Sorbitol.
Biochem/physiol actions: activity of sorbitol dehydrogenase increases in diabetic animals.
Unit definition: one unit will convert 1,0 µmole of D-fructose to D-sorbitol per minute at pH 7,6 at 25 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge