Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Eosin Y, orangenes Pulver C.I. 45380:2

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0215792205 0215792225
ICNA0215792205EA 72.7 EUR InStock
ICNA0215792205 ICNA0215792225
Eosin Y, orangenes Pulver C.I. 45380:2
Eosin Y
Eosin Y is a counterstain for hematoxylin.

It is used in combination with Azure II to show morphological details of marrow cells; buffered Azure A/Eosin Y method for demostrating cell granules, microorganisms and nuclei; for showing insect musculature; for demonstrating cells infected with mosquito-iridescent virus; use in combination with Alum Hematoxylin/Azure II to show mast cells, early necrobiotic changes in cells and protozoans, rickettsias and bacteria in tissues; to demonstrate volutin granules in any microorganism; bond's modification of mann's eosin-methyl blue for negri bodies in smears; harris stain for negri bodies in smears; mann's method for staining negri bodies in sections; gridley's stain for endamoeba in tissue sections; a modified methenamine-silver nitrate method for the demonstration of endamoeba in sections.

Soluble in water (0,8 mg/ml); ethanol (10 mg/ml); ethylene glycol monomethyl ether (methyl-cellosolve; EGME) (40 mg/ml).


Formel: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
Melting Pt: 295
MDL Number: MFCD00036189
CAS-Nummer: 15086-94-9
Order Now
Spezifikation Testergebnisse
Identity Test Passes

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge