Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Xylazine hydrochloride, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

0215830725 0215830701 0215830705
ICNA0215830725EA 1107 EUR InStock
ICNA0215830725 ICNA0215830701 ICNA0215830705
Xylazine hydrochloride, weißes Pulver
Xylazine hydrochloride
Xylazine hydrochloride is an alpha agonist with sedative, analgesic and muscle relaxant properties.

  • Presentation : White Powder
  • Solubility : Soluble in water (50 mg/mL).
  • Key Applications : To produce rapid and reversible anesthesia in experimental animals
  • Product Type : Biochemicals

A solution of ketamine hydrochloride and xylazine hydrochloride has been used together to produce rapid and reversible anesthesia in experimental animals. Xylazine elicited a diuretic response in ketamine-anesthesised animals, probably through activation of complex peripheral and CNS α2-adrenergic receptor systems.

α2-Adrenergic receptor agonist, sedative, muscle relaxant.


Formel: C₁₂H₁₆N₂S
Melting Pt: 161,..171 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00058196
CAS-Nummer: 23076-35-9
UN: 2811
ADR: ,III
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
Melting Point 161 - 171°C

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge