Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Lecithin aus Eigelb 96, powder

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


02160059.2 02160059.5 0216005905 02160059.1 0216005901
ICNA02160059.2EA 152 EUR InStock
ICNA02160059.2 ICNA02160059.5 ICNA0216005905 ICNA02160059.1 ICNA0216005901
Lecithin aus Eigelb 96, powder
Lecithin aus Eigelb
Lecithin is the major membrane phospholipid in eukaryotic cells. It also serves as a reservoir for several lipid messengers.

Lecithin is used for preparation of vesicle suspensions commonly called liposomes or as monolayers. It is the source of the bioactive lipids lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, plateletactivating factor, and arachidonic acid.

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.

Soluble in fat, paraffin, ether, ethanol, chlorinated hydrocarbons(100 mg/ml, CHCl3). Dispersible in water.


Storage Temperature: Kühlschrank
MDL Number: MFCD00131449
CAS-Nummer: 93685-90-6
EINECS: 297-639-2
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge