Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

β-Cyclodextrin, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219005305 0219005325 0219005380
ICNA0219005305EA 32.4 EUR InStock
ICNA0219005305 ICNA0219005325 ICNA0219005380
β-Cyclodextrin, weißes Pulver
β-Cyclodextrin
β-cyclodextrin is most commonly used as a complexing agent. It is reported to be useful for the selective precipitation of enantiomeric, positional or structural isomers. It also forms a water soluble complex with dansyl chloride for fluorescent labeling of proteins.

Cyclodextrins are cyclic oligosaccharides consisting of 6, 7, or 8 glucopyranose units with hydrophobic interiors, usually referred to as α-, β-, or γ-cyclodextrins, respectively. Lipophilic drugs of a size compatible with the hydrophobic core of a cyclodextrin can form complexes, resulting in increased aqueous solubility of the drugs. Cyclodextrins are non-toxic in many species (mice and rabbits), and do not denature proteins or interfere with enzymatic reactions.

Soluble in water, 1 M NH4OH: 50 mg/ml (clear, colorless solution).


Formel: C₄₂H₇₀O₃₅
MW: 1134.98 g/mol
Melting Pt: 298…300 °C (Lit.)
Dichte: 1.624 g/cm³ (Lit.)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00078139
CAS-Nummer: 7585-39-9
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 5 - 7.5
Loss on Drying ≤14%
Optical Rotation 158 ± 6° (c=1,water)

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge