Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Erythromycin ≥98%, Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219019701 0219019705
ICNA0219019701EA 24.4 EUR InStock
ICNA0219019701 ICNA0219019705
Erythromycin ≥98%, Pulver
Erythromycin
Erythromycin is a macrolide antibiotic that bind to the 50S ribosomal subunit and inhibit protein synthesis. This is an antimicrobial substance produced by the growth of certain strains of Streptomyces erythreus. It is a mixture consisting largely of erythromycin A with lesser amounts of erythromycins B and C.

Used in cell culture applications at approximately 100 mg/liter. Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth. Erythromycin resistance can be induced in bacteria. Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth.

Mode of Action: Inhibits protein synthesis (elongation) at the level of transpeptidation (aminoacyl translocation A-site to P-site).
Antimicrobial spectrum: Gram-negative and gram-positive bacteria.

Slightly soluble in water (~2 mg/ml); moderately soluble in ether, ethylene dichloride, amyl acetate; freely soluble in alcohols (ethanol - 50 mg/ml), acetone, chloroform, acetonitrile, ethyl acetate. Soluble in 2 M Hydrochloric acid (50 mg/ml).


Formel: C₃₇H₆₇NO₁₃
MW: 733.95 g/mol
MDL Number: MFCD00084654
CAS-Nummer: 114-07-8
Order Now
Spezifikation Testergebnisse
Synonyms E- mycin; Erycin; Robimysin; Erythrocin
Form White Powder
Storage Store at Room Temperature (15 - 30 °C)
Extinction Coefficient EM = 50 (280 nm, pH 6,3)
Moisture Content 10%
Optical Rotation -71° to -78° (c=2, ethanol)
pH 8,0 - 10,5 (70% aq soln)
pka 8,8
Potency ≥850 μg/mg

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge