Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Lysin Monohydrat, powder

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219022405 0219022425 0219022480
ICNA0219022405EA 50.7 EUR InStock
ICNA0219022405 ICNA0219022425 ICNA0219022480
L(+)-Lysin Monohydrat, powder
L(+)-Lysin Monohydrat
L-Lysine is an essential proteinogenic α amino acid. It contains an N-butyl amino group in the side chain, and this moiety is protonated at physiological pH. In addition, L-lysine is one of the two amino acids that are degraded to give ketone bodies (a ketogenic amino acid).

L-Lysine is used in a wide range of applications including as a supplement in cell culture media; a substrate of enzymes such as L-lysine oxidase (EC 1.4.3.14); a component of poly-lysine polymers; and a substrate for oxidation and glycation mechanism studies. L-lysine has been used with cultured human osteoblasts to study cell proliferation.

Lysine is degraded in vivo to give acetoacetyl CoA via an initial transamination with alpha-ketoglutarate. L-Lysine is an essential proteinogenic α amino acid used in a wide range of applications including as a supplement in cell culture media; a substrate of enzymes such as L-lysine oxidase (EC 1.4.3.14); a component of poly-lysine polymers; and a substrate for oxidation and glycation mechanism studies.

Soluble in 0,1 N hydrochloride (50 mg/ ml - clear, colorless solution) or water (100 mg/ml - clear, colorless solution).


Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MW: 164.2 g/mol
Melting Pt: 225
MDL Number: MFCD00151035
CAS-Nummer: 39665-12-8
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 9.9 - 10.3
Loss on Drying ≤11.8%
Optical Rotation (c=8, 6 N HCl) +23.5° to +26.5°

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge