Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat ≥98%, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0219032025 02190320.5 0219032001 0219032005
ICNA0219032025EA 2217 EUR InStock
ICNA0219032025 ICNA02190320.5 ICNA0219032001 ICNA0219032005
3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat ≥98%, weißes Pulver
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
CHAPSO is the hydroxy analog of CHAPS; a non denaturing zwitterionic detergent for solubilising proteins, especially integral membrane proteins while maintaining the functional capability of the protein of interest.

  • Useful for the solubilisation of integral membrane proteins
  • Critical micelle concentration (CMC): 8 mM
  • Aggregation Number:11

It has a more polar head group than CHAPS, giving it a greater solubility. CHAPSO has a high CMC (8 mM) and does not denature membrane-bound proteins. It can solubilise the opiate receptor in a state exhibiting reversible binding of opiates.

CHAPSO is the hydroxy analog of CHAPS; a nondenaturing zwitterionic detergent for solubilising proteins, especially integral membrane proteins while maintaining the functional capability of the protein of interest. It has a more polar head group than CHAPS, giving it a greater solubility. CHAPSO has a high CMC (8 mM) and does not denature membrane-bound proteins. It can solubilise the opiate receptor in a state exhibiting reversible binding of opiates.

Soluble in water (100 mg/ml - clear, colourless to slight yellowish solution), methanol (40 mg/ml- clear, colourless solution).


Formel: C₃₂H₅₈N₂O₈S
MW: 630.89 g/mol
Melting Pt: 184…186 °C (Lit.)
Storage Temperature: Raumtemperatur
CAS-Nummer: 82473-24-3
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (10% aq soln) 5,0 - 9,0
OD350 nm (1% aq soln) <0,04

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge