Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Folinic acid calcium salt pentahydrate, blassgelbes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219150425 0219150480 0219150490 0219150491
ICNA0219150425EA 53 EUR InStock
ICNA0219150425 ICNA0219150480 ICNA0219150490 ICNA0219150491
Folinic acid calcium salt pentahydrate, blassgelbes Pulver
Folinic acid calcium salt pentahydrate
Folinic acid calcium salt, pentahydrate offsets the effects of folic acid antagonists that act by binding dihydrofolate reductase and blocking the conversion of folic acid to tetrahydrofolate.

In medicine, calcium folinate is used after treatment with methotrexate to reduce the toxicity of the methotrexate. Calcium folinate is also used to enhance the cytotoxic effect of 5-fluorouracil in the treatment of cancer.

Intermediate product of the metabolism of folic acid; the active form into which that acid is converted in the body, ascorbic acid being a necessary factor in the conversion process.

Freely soluble in water (100 mg/mL-clear to slightly hazy solution); almost insoluble in ethanol, alcohol.


Formel: C₂₀H₃₁CaN₇O₁₂
MDL Number: MFCD00149465
CAS-Nummer: 6035-45-6
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge