Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid ≥98%, gelbes Pulver Reagenz für die Zellkultur

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0219459291 0219459280 0219459290
ICNA0219459291EA 104 EUR InStock
ICNA0219459291 ICNA0219459280 ICNA0219459290
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid ≥98%, gelbes Pulver Reagenz für die Zellkultur
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Storage: +4°C, protect from light
3- (4,5-Dimethyl-2-thiazolyl-2)-2,5-diphenyl-tetrazolium bromide is used as a colorimetric metabolic activity indicator in cell viability assays. Thiazolyl Blue Tetrazolium Blue (MTT) is a dye used in measurement of cell proliferation. MTT produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells. The blue crystals are solubilized with acidified isopropanol and the intensity is measured colorimetrically at 570 nm. MTT has been used as a histochemical/cytochemical reagent and for the detection of NAD. ADP-linked enzyme systems in tissue cannot be detected with MTT, due to binding of the cation by the cyanide trap used. MTT is rapidly reduced to the formazan, which chelates with nickel, copper, and cobalt; the cobalt chelate has been used in oxidative systems.

  • Cell Culture Reagent Grade
  • Presentation: Yellow Powder
  • Soluble in methyl alcohol (heat), methanol (10 mg/mL with heat - clear, yellow solution)and water (5 mg/mL-clear to very slightly hazy, yellow to bright yellow solution).


Formel: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Melting Pt: 189…199 °C
Storage Temperature: Kühlschrank
MDL Number: MFCD00011964
CAS-Nummer: 298-93-1
EINECS: 206-069-5
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Purity ~98%
FTIR Conforms to Standard
Melting Point 189 - 199°C
UV/Vis: I max (methanol) 376 ± 2 nm; 242 ± 2 nm

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge