Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Trihydrat ≥99%, weißes Pulver Reagenz für die Zellkultur

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219461310 0219461301 0219461305
ICNA0219461310EA 280 EUR InStock
ICNA0219461310 ICNA0219461301 ICNA0219461305
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Trihydrat ≥99%, weißes Pulver Reagenz für die Zellkultur
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Trihydrat
Storage: -20 °C.
Adenosine 5'-triphosphate (ATP) and its phosphate bonds are the basic components of energy exchange in many biological systems.
Adenosine 5'-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is used in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps. ATP serves as a coenzyme in a wide array of enzymatic reactions.
P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems.

  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Extinction Coefficient (E1%): 15.4 (259 nm in 100 mM phosphate buffer, pH 7.0). (Lit.)
  • pKa: 4.0 (amino group); 6.5 (secondary phosphate) (Lit.)


Formel: C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
MW: 605.19 g/mol
Melting Pt: 185…190 °C (Lit.)
Storage Temperature: Gefrierschrank
MDL Number: MFCD00150754
CAS-Nummer: 51963-61-2
EINECS: 213-579-1
Order Now
Spezifikation Testergebnisse
Identity Test A Passes
Identity Test B Passes
Identity Test C Passes
Purity ~ 99%
FTIR Conforms to Standard
I max (H20) 258 ± 2 nm
OD250/OD259 0.75-0.83
pH (5% aq soln) Acidic to Litmus Paper
Solubility (5% aq soln) Clear to Slightly Hazy Solution

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge