Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Asparagin, wasserfrei ≥99%, weißes Pulver Reagenz für die Zellkultur

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219463025 0219463080 0219463090
ICNA0219463025EA 49.2 EUR InStock
ICNA0219463025 ICNA0219463080 ICNA0219463090
L(+)-Asparagin, wasserfrei ≥99%, weißes Pulver Reagenz für die Zellkultur
L(+)-Asparagin
Storage: Store at Room Temperature (15-30 °C). Store Desiccated.
L-Asparagine is used in cell culture media and is a component of MEM non-essential amino acids solution. L-Asparagine has been shown to enhance ornithine decarboxylase activity in cultured human colon adenocarcinoma Caco-2 cells and in cultured IEC-6 intestinal epithelial cells. Spore germination in Bacillus subtilis has been increased in the presence of L-asparagine. An isoxazoline RGD mimic platelet GPIIb/IIIa antagonist has been prepared by chiral synthesis with L-asparagine as a starting material. L-Asparagine has been utilized in the synthesis of 4-azalysine building blocks for application to combinatorial chemistry.

  • Anhydrous
  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • pKa: 2.14 (-COOH), 8.72 (-NH2) (Lit.)
  • Soluble in aqueous solution (1%-clear, colorless solution).


Formel: H₂NCOCH₂CH(NH₂)CO₂H
Melting Pt: 234…235 °C (Lit.)
MDL Number: MFCD00064401
CAS-Nummer: 70-47-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge