Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Kaliumdihydrogenphosphat, wasserfrei, weißes kristallines Pulver Reagenz für die Zellkultur

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0219472780 0219472790 0219472791
ICNA0219472780EA 26.8 EUR InStock
ICNA0219472780 ICNA0219472790 ICNA0219472791
Kaliumdihydrogenphosphat, wasserfrei, weißes kristallines Pulver Reagenz für die Zellkultur
Kaliumdihydrogenphosphat
Storage: Room Temperature, desiccate
Potassium phosphate is a reagent with high buffering capacity. It occurs in several forms: monobasic, dibasic and tribasic (K3PO4). Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.
Potassium phosphate monobasic is typically used as a component for a wide variety of media used in the culture of microorganisms. It is widely used in molecular biology, biochemistry, and chromatography. It is a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.

  • Anhydrous
  • Cell Culture Reagent Grade
  • Presentation: White Crystalline Powder
  • pKa: (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid) at 25 °C (Lit.)
  • Soluble in water (100 mg/mL); Insoluble to slightly soluble in ethanol; insoluble in alcohol.


Formel: KH₂PO₄
MW: 136.09 g/mol
Siedepunkt: 100
Melting Pt: 250 °C (Zers., Lit.)
Dichte: 2.338 g/mL (25 °C) (Lit.)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011401
CAS-Nummer: 7778-77-0
Merck Index: 13,07744
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Assay ≥99.0%
pH (5% aq soln) 4.1-4.5

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge