Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Protamine sulphate (aus Lachssperma), ≥100 USP u/mg, weißes Pulver Reagenz für die Zellkultur

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219472925 0219472980 0219472901 0219472905
ICNA0219472925EA 242 EUR InStock
ICNA0219472925 ICNA0219472980 ICNA0219472901 ICNA0219472905
Protamine sulphate (aus Lachssperma), ≥100 USP u/mg, weißes Pulver Reagenz für die Zellkultur
Protamine sulphate
Storage: Room Temperature, desiccate
Protamine sulfate is a purified mixture of simple protein principles obtained from the sperm or testes of suitable species of fish, which has the property of neutralizing heparin. Because of having many basic amino acids (mostly arginine) protamine contains far greater nitrogen than other proteins. Its molecular weight is relatively small. Histone and other basic proteins in the testes of unmatured fishes convert into protamine along with the growth of the fishes. In the testes, protamine takes the form of nucleoprotamine linked with DNA.
Protamine Sulfate is a raw material for study preparations like insulin compounds, and etc. It is used in separation and refining of vaccines. It is a reagent for removing nucleic acids from enzyme solution for the purpose of easy separation and refining. Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.
Protamine sulfate is a small cationic protein that binds and precipitates DNA. Inhibits lipoprotein lipase. Protamine sulfate shown to inhibit the classical pathway of complement. It inhibits turnover of lipoproteins by lipoprotein lipase.

  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Freely soluble in hot water, slightly soluble in cold water; soluble in 0.1 M KH2PO4; Soluble in HCl. Practically insoluble in organic solvents.


Storage Temperature: Raumtemperatur
MDL Number: MFCD00132091
CAS-Nummer: 9009-65-8
Order Now
Spezifikation Testergebnisse
Identity Test Passes
Source Salmon Sperm
Loss on Drying ≤5%
Sulfate 16-22%
Nitrogen 22.5-25.5%
Activity ≥100 USP u/mg

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge